Skip to Content
Merck
All Photos(1)

Key Documents

740365

Sigma-Aldrich

cis-4-Amino-cyclohexanol hydrochloride

97%

Synonym(s):

cis-4-Hydroxycyclohexylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14ClNO
Molecular Weight:
151.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

193-200 °C

storage temp.

2-8°C

SMILES string

Cl.N[C@@H]1CC[C@H](O)CC1

InChI

1S/C6H13NO.ClH/c7-5-1-3-6(8)4-2-5;/h5-6,8H,1-4,7H2;1H/t5-,6+;

InChI key

RKTQEVMZBCBOSB-KNCHESJLSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service