Skip to Content
Merck
All Photos(1)

Key Documents

739456

Sigma-Aldrich

Poly(ethylene glycol) methyl ether 2-bromoisobutyrate

average Mn 1,200

Synonym(s):

PEG methyl ether bromoisobutyrate 1000, mPEG bromoisobutyrate 1000, ATRP initiator, PEG methyl ether bromoisobutyrate, mPEG bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCOC(CH3)2Br
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 1,200

transition temp

Tm 35-40 °C

Looking for similar products? Visit Product Comparison Guide

General description

PEG average Mn = 1,000; n ~22

Application

Poly(ethylene glycol)-containing ATRP initiator for generating a block copolymer with a PEG block on one end. The PEG block is terminated with an unreactive methoxy group.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Protocols

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service