Skip to Content
Merck
All Photos(1)

Key Documents

556122

Sigma-Aldrich

4-Chlorobenzyl acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2OCOCH3
CAS Number:
Molecular Weight:
184.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5200 (lit.)

bp

78-81 °C/1 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCc1ccc(Cl)cc1

InChI

1S/C9H9ClO2/c1-7(11)12-6-8-2-4-9(10)5-3-8/h2-5H,6H2,1H3

InChI key

MAGQINDMZBNWLG-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition-Metal-Catalyzed Benzylation of Arenes and Heteroarenes.
Mertins K, et al.
Angewandte Chemie (International Edition in English), 44(2), 238-242 (2005)
Bismuth (III) salts as convenient and efficient catalysts for the selective acetylation and benzoylation of alcohols and phenols.
Mohammadpoor-Baltork I, et al.
Tetrahedron, 57(27), 5851-5854 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service