About This Item
Recommended Products
Assay
95%
refractive index
n20/D 1.404 (lit.)
bp
83 °C (lit.)
density
1.393 g/mL at 25 °C (lit.)
SMILES string
Fc1cc(F)c(F)c(F)c1
InChI
1S/C6H2F4/c7-3-1-4(8)6(10)5(9)2-3/h1-2H
InChI key
UHHYOKRQTQBKSB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service