Skip to Content
Merck
All Photos(1)

Key Documents

292990

Sigma-Aldrich

S-Furfuryl thioacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

99%

refractive index

n20/D 1.526 (lit.)

bp

90-92 °C/12 mmHg (lit.)

density

1.171 g/mL at 25 °C (lit.)

SMILES string

CC(SCC1=CC=CO1)=O

InChI

1S/C7H8O2S/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

LQOUTUIIYXYBQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

S-Furfuryl thioacetate (S-2-furfuryl thioacetate) has been used in lipase (from Candida rugosa) mediated synthesis of 2-furfurylthiol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachid Bel Rhlid et al.
Journal of agricultural and food chemistry, 50(14), 4087-4090 (2002-06-27)
Enzymatic hydrolysis of S-3-(2-methylfuryl) thioacetate and S-2-furfuryl thioacetate using lipase from Candida rugosa produced 2-methyl-3-furanthiol and 2-furfurylthiol, respectively. When reactions were carried out at room temperature and pH 5.8, 2-methyl-3-furanthiol was produced in a optimal yield of 88% after 15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service