Skip to Content
Merck
All Photos(1)

Key Documents

258288

Sigma-Aldrich

Chlorophyllin sodium copper salt

Synonym(s):

Chlorophyllin, coppered trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H31CuN4Na3O6
CAS Number:
Molecular Weight:
724.15
EC Number:
UNSPSC Code:
12352103

SMILES string

[Na+].[Na+].[Na+].CCc1c(C)c2cc3c(C=C)c(C)c4cc5nc(C(CCC([O-])=O)C5C)c(CC([O-])=O)c6c(C([O-])=O)c(C)c(cc1n2)n6[Cu]n34

InChI

1S/C34H36N4O6.Cu.3Na/c1-7-19-15(3)23-12-25-17(5)21(9-10-29(39)40)32(37-25)22(11-30(41)42)33-31(34(43)44)18(6)26(38-33)14-28-20(8-2)16(4)24(36-28)13-27(19)35-23;;;;/h7,12-14,17,21H,1,8-11H2,2-6H3,(H5,35,36,37,38,39,40,41,42,43,44);;;;/q;+2;3*+1/p-5

InChI key

HWDGVJUIHRPKFR-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoungju Choi et al.
Toxicology letters, 241, 49-59 (2015-11-26)
In vitro cell culture systems are a useful tool to rapidly assess the potential safety or toxicity of chemical constituents of food. Here, we investigated oxidative stress and organ-specific antioxidant responses by 7 potential dietary ingredients using canine in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service