Skip to Content
Merck
All Photos(1)

Key Documents

197610

Sigma-Aldrich

2-(3-Bromophenyl)-1,3-dioxolane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BrO2
CAS Number:
Molecular Weight:
229.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.563 (lit.)

bp

132-133 °C/8 mmHg (lit.)

density

1.514 g/mL at 25 °C (lit.)

SMILES string

Brc1cccc(c1)C2OCCO2

InChI

1S/C9H9BrO2/c10-8-3-1-2-7(6-8)9-11-4-5-12-9/h1-3,6,9H,4-5H2

InChI key

VYPYKCPWNPPBBX-UHFFFAOYSA-N

Application

2-(3-Bromophenyl)-1,3-dioxolane was used in the synthesis of acetal by arylation of 1-benzylpiperidin-4-amine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew M Griffin et al.
Bioorganic & medicinal chemistry letters, 19(21), 5999-6003 (2009-10-06)
We have investigated phenol replacements in a series of diaryl amino piperidine delta opioid agonists. From this study we have demonstrated that the hydroxy functional group can be replaced with a primary amide group, giving enhanced activity at the delta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service