Skip to Content
Merck
All Photos(1)

Documents

186104

Sigma-Aldrich

3,5-Dimethylpiperidine, mixture of cis and trans

≥99%

Synonym(s):

3,5-Lupetidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

liquid

refractive index

n20/D 1.4454 (lit.)

bp

144 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

SMILES string

CC1CNCC(C)C1

InChI

1S/C7H15N/c1-6-3-7(2)5-8-4-6/h6-8H,3-5H2,1-2H3

InChI key

IDWRJRPUIXRFRX-UHFFFAOYSA-N

Application

3,5-Dimethylpiperidine was used to investigate the influence of substituents on heterocyclic piperidine as acid corrosion inhibitors for iron. It was used in the synthesis of pure phase ZSM-11 using extremely dense gels with minimal water contents.
Reactant for synthesis of:
Substituted bisphenol A derivatives as β-amyloid peptide aggregation inhibitors
MMP-13 selective isonipecotamide α-sulfone hydroxamates
Gem-diamines for active organocatalysts for biodiesel production
Benzimidazole inhibitors of the antigen receptor-mediated NF-κB

Reactant for Mannich reactions to form Ru(II) complexes

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple, efficient syntheses of zeolite ZSM-11 (MEL) at temperatures below 100? C using very dense gels.
Fyfe CA, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 150, 7-13 (2012)
Piperidines as corrosion inhibitors for iron in hydrochloric acid.
Khaled KF, et al.
J. Appl. Electrochem., 34(7), 697-704 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service