Skip to Content
Merck
All Photos(1)

Key Documents

157651

Sigma-Aldrich

Sodium 2-chloroethanesulfonate monohydrate

98%

Synonym(s):

2-Chloroethanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2SO3Na · H2O
CAS Number:
Molecular Weight:
184.57
Beilstein:
5649202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

292 °C (dec.) (lit.)

functional group

chloro
sulfonic acid

SMILES string

[Na+].[H]O[H].[O-]S(=O)(=O)CCCl

InChI

1S/C2H5ClO3S.Na.H2O/c3-1-2-7(4,5)6;;/h1-2H2,(H,4,5,6);;1H2/q;+1;/p-1

InChI key

OSEJMOGPHHVJJG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 2-chloroethanesulfonate monohydrate was used as sulfoethylating agent in the preparation of sulfoethyl cellulose.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of low sulfoethylated cellulose.
Zhang K, et al.
Carbohydrate Polymers, 83(2), 616-622 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service