Skip to Content
Merck
All Photos(1)

Key Documents

122025

Sigma-Aldrich

3-Ethyl-2-thioxo-4-oxazolidinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2S
CAS Number:
Molecular Weight:
145.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

39-41 °C (lit.)

functional group

thiocarbamate

SMILES string

CCN1C(=O)COC1=S

InChI

1S/C5H7NO2S/c1-2-6-4(7)3-8-5(6)9/h2-3H2,1H3

InChI key

ZILKBTSQUZJHOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Ethyl-2-thioxo-4-oxazolidinone may be used in chemical synthesis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service