Enzymatic Assay: Glucose Oxidase
1. Objective
The objective of this procedure is to standardize a procedure for the enzymatic assay of glucose oxidase.
2. Scope
This procedure applies to all products that have a specification for glucose oxidase activity.
3. Definitions
- Purified Water - water from a deionizing system, resistivity > or = 18MΩ•cm at 25 °C
- GO = Glucose Oxidase
- POD = Peroxidase
- Unit definition of glucose oxidase = One unit will oxidize 1.0 μMol of β-D-glucose to D-gluconolactone and H2O2 per minute at pH 5.1 at 35 °C (equivalent to an O2 uptake of 22.4 μL per minute). If the reaction mix is saturated with oxygen, the activity may increase by up to 100%.
4. Discussion
- β -D-Glucose + O2 + H2O GO > D-Glucono -1,5 – Lactone + H2O2
- H2O2 + o-Dianisidine(reduced) POD > o-Dianisidine(oxidized)
5. Responsibilities
Analytical Services personnel should follow this protocol as written.
6. Safety
Refer to the Safety Data Sheet (SDS) for hazards and appropriate handling precautions.
7. Procedure
Conditions
T=35 °C, pH= 5.1, A , Light path=1 cm
Method
Continuous Spectrophotometric Rate Determination
Reagents
- 50 mM Sodium Acetate Buffer, pH 5.1 at 35 °C (Buffer): Prepare 500 mL in purified water saturated with oxygen (saturate the water for five minutes and let it stand overnight before running the assay) using sodium acetate, trihydrate, (Product No. S8625). Adjust to pH 5.1 at 35 °C with 1 M HCL.)
- 0.21 mM o-dianisidine Solution (o-DDH): Dissolve 20 mg of o-dianisidine dihydrochloride (Product No. D9154) , or (Product No. F5803) , in 8 mL purified water saturated in oxygen using an amber vial or other vial protected from light. Then dilute 5.35 mL to 200 mL with 50 mM Sodium Acetate Buffer, pH 5.1 at 35 °C (Buffer).
- 10% (w/v) β-D-(+) Glucose Substrate Solution: Prepare 60 mL in purified water using β-D-(+) Glucose.
- 0.17 mM o-dianisidine and 1.72% (w/v) glucose solution (Reaction Cocktail): Immediately before use, prepare 232 mL by combining 192 mL of 0.21 mM o-dianisidine Solution (o-DDH) with 40 mL of 10% (w/v) β-D-(+) Glucose Substrate Solution. Equilibrate to 35 °C and adjust to pH 5.1 if necessary with 1 M HCL. Prepare fresh.
- Peroxidase Enzyme solution (POD): Immediately before use, prepare a solution containing 60 purpurogallin units/mL of POD Type II in cold purified water.
- Glucose Oxidase Enzyme Solution (GO): For all Glucose Oxidase products (except for crude and liquid products) prepare an initial solution of 20-40 units/mL in cold buffer, and immediately before use, dilute to 0.4-0.8 units/mL in cold buffer (this is zero time). Keep the initial solution on ice. Make a fresh second dilution in cold buffer and run again after 60 minutes (this is 60 min time). For all crude and liquid products see table below.
Crude products and liquids | First Dilution | Second Dilution |
---|---|---|
G1262 | 0.2 mg/mL | No further dilution required |
G6766 | 0.2 mg/mL | No further dilution required |
G6125 | 2.0 mg/mL | 0.1 mL+6.4 mL Buffer |
G6641 | 1.0 mg/mL | 0.1 mL+5.0 mL Buffer |
G9010 | 0.1 mL+5.0 mL Buffer | 0.1 ml+3.0 mL Buffer |
G2133 | 0.1 mg/mL | 0.1 mL+5.0 mL Buffer |
G7016 | 0.2 mg/mL | 0.1 mL+5.0 mL Buffer |
G7141 | 0.1 mg/mL | 0.1 mL+5.0 mL Buffer |
G7779 (1.0 mg/mL) | 0.1 mL+0.5 mL Buffer | 0.1 mL+5.0 mL Buffer |
G7779 (0.01 mg /mL) | Neat | 1.0 mL+3.0 mL Buffer |
Test Method
Pipette the following reagents into suitable cuvettes:
Test | Blank | |
---|---|---|
0.17 mM o-dianisidine and 1.72% (w/v) glucose solution (Reaction Cocktail) | 2.9 | 2.9 |
Peroxidase Enzyme solution (POD) | 0.1 | 0.1 |
Then add:
Glucose Oxidase Enzyme Solution (GO) | 0.1 | ----- |
50 mM Sodium Acetate Buffer, pH 5.1 at 35 °C (Buffer) | ----- | 0.1 |
Immediately mix by inversion and record the increase in A500nm/min for six minutes. Obtain the maximum linear rate for both the Test and the Blank using a minimum of a 1.0 minute period and a minimum of four A500nm points.
Calculations
Units/mL enzyme = | (ΔA500nm/minTest - ΔA500nm/ min Blank) * (3.1) * (df) |
(7.5) * (0.1) |
3.1=volume (in milliliters) of assay
df=Dilution factor
7.5=Millimolar extinction coefficient of oxidized o-dianisidine at 500 nM
0.1=Volume (in milliliters) of enzyme used
Units/mg solid = | Units/mL enzyme | |
mg solid/mL enzyme |
Units/mg protein = | Units/mL enzyme | |
mg protein/mL enzyme |
Final Assay Concentration
In a 3.10 mL reaction mix, the final concentrations are 48 mM sodium acetate, 0.16 mM o-dianisidine, 1.61% (w/v) glucose, and 6 units peroxidase (concentration of glucose oxidase will vary as to which product is used.)
Network error: Failed to fetch