Merck
  • JNK regulates APP cleavage and degradation in a model of Alzheimer's disease.
JNK regulates APP cleavage and degradation in a model of Alzheimer's disease.
Neurobiology of disease (2009-01-27)
Alessio Colombo, Antonio Bastone, Cristina Ploia, Alessandra Sclip, Mario Salmona, Gianluigi Forloni, Tiziana Borsello
ABSTRACT
Secretion of Amyloid-beta peptide (Abeta) circulating oligomers and their aggregate forms derived by processing of beta-amyloid precursor protein (APP) are a key event in Alzheimer's disease (AD). We show that phosphorylation of APP on threonine 668 may play a role in APP metabolism in H4-APP(sw) cell line, a degenerative AD model. We proved that JNK plays a fundamental role in this phosphorylation since its specific inhibition, with the JNK inhibitor peptide (D-JNKI1), induced APP degradation and prevented APP phosphorylation at T668. This results in a significant drop of betaAPPs, Abeta fragments and Abeta circulating oligomers. Moreover the D-JNKI1 treatment produced a switch in the APP metabolism, since the peptide reduced the rate of the amyloidogenic processing in favour of the non-amyloidogenic one. All together our results suggest an important link between APP metabolism and the JNK pathway and contribute to shed light on the molecular signalling pathway of this disease indicating JNK as an innovative target for AD therapy.
Sigma-Aldrich
Hygromycin B from Streptomyces hygroscopicus, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.