Merck
  • Novel generation of deep eutectic solvent as an acceptor phase in three-phase hollow fiber liquid phase microextraction for extraction and preconcentration of steroidal hormones from biological fluids.
Novel generation of deep eutectic solvent as an acceptor phase in three-phase hollow fiber liquid phase microextraction for extraction and preconcentration of steroidal hormones from biological fluids.
Talanta (2017-11-16)
Mohammad Mahdi Khataei, Yadollah Yamini, Ali Nazaripour, Meghdad Karimi
ABSTRACT
In this study, a novel generation of deep eutectic solvents (DESs) was used as an acceptor phase in three-phase hollow fiber liquid phase microextraction (HF-LPME) based on two immiscible organic phases. It was compared with other common DESs for extraction and preconcentration of dydrogesterone (DYD) and cyproterone acetate (CPA) from urine and plasma samples. The extracted analytes were analyzed by high performance liquid chromatography with UV-vis detector (HPLC-UV). This phosphonium based DES due to low volatility, low price and multifunctionality introduced itself as worthy next generation of acceptor phase in HF-LPME. The factors affected on extraction efficiency of the analytes were investigated and optimized. The performance of the proposed method was studied in terms of linear ranges (LRs from 1 to 500µgL
Sigma-Aldrich
Methyltriphenylphosphonium iodide, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyltriphenylphosphonium bromide, 98%
Pricing and availability is not currently available.