- Identification of a selective small-molecule inhibitor of type 1 adenylyl cyclase activity with analgesic properties.
Identification of a selective small-molecule inhibitor of type 1 adenylyl cyclase activity with analgesic properties.
Adenylyl cyclase 1 (AC1) belongs to a group of adenylyl cyclases (ACs) that are stimulated by calcium in a calmodulin-dependent manner. Studies with AC1 knockout mice suggest that inhibitors of AC1 may be useful for treating pain and opioid dependence. However, nonselective inhibition of AC isoforms could result in substantial adverse effects. We used chemical library screening to identify a selective AC1 inhibitor with a chromone core structure that may represent a new analgesic agent. After demonstrating that the compound (ST034307) inhibited Ca