Merck
  • Pd(0)-Catalyzed carbonylation of 1,1-dichloro-1-alkenes, a new selective access to Z-α-chloroacrylates.
Pd(0)-Catalyzed carbonylation of 1,1-dichloro-1-alkenes, a new selective access to Z-α-chloroacrylates.
Chemical communications (Cambridge, England) (2010-09-21)
Martin Arthuis, Anne Lecup, Emmanuel Roulland
ABSTRACT
A novel and fully chemo- and stereoselective three component strategy leading to Z-α-chloroacrylates by a Pd(0)-catalyzed reaction of CO (1 atm) with 1,1-dichloro-1-alkenes and various alcohols is disclosed. This catalytic approach compares favourably with the Wittig type strategies as α-chloroacrylates of pure Z configuration are obtained in high yield.
Sigma-Aldrich
Tris(dibenzylideneacetone)dipalladium(0), 97%
Pricing and availability is not currently available.