Merck
  • Influence of different feeding regimes on the survival, growth, and biochemical composition of Acropora coral recruits.
Influence of different feeding regimes on the survival, growth, and biochemical composition of Acropora coral recruits.
PloS one (2017-11-29)
Jessica A Conlan, Craig A Humphrey, Andrea Severati, David S Francis
ABSTRACT
Heterotrophic feeding in newly-settled coral planulae can potentially improve survivorship and accelerate early development in some species; however, an optimal diet to facilitate this does not currently exist. This study evaluated the efficacy of three heterotrophic feeding regimes (enriched rotifers, unfiltered seawater, and a novel, particulate diet), against a wholly-phototrophic treatment on Acropora hyacinthus, A. loripes, A. millepora, and A. tenuis recruits, over 93 days post-settlement. The unfiltered seawater treatment recorded maximum survival for all species (A. hyacinthus 95.9±8.0%, A. loripes: 74.3±11.5%, A. millepora: 67±12.7%, A. tenuis: 53.2±11.3%), although not significant. Growth (% surface area gain) was also greatest in the unfiltered seawater, and this was significant for A. millepora (870±307%) and A. tenuis (693±91.8%) (p<0.05). Although total lipid concentration was relatively stable across treatments, the lipid class composition exhibited species-specific responses to each treatment. Lower saturated and higher polyunsaturated fatty acids appeared beneficial to recruit performance, particularly in the unfiltered seawater, which generally contained the highest levels of 20:5n-3 (EPA), 22:6n-3 (DHA), and 20:4n-6 (ARA). The present study demonstrates the capacity of a nutritionally adequate and readily accepted heterotrophic feeding regime to increase coral recruit survival, growth, and health, which can greatly reduce the time required in cost- and labour-intensive culture.
Sigma-Aldrich
DL-α-Palmitin, ≥99%
Pricing and availability is not currently available.