Merck
  • Ultra-high mechanical properties of porous composites based on regenerated cellulose and cross-linked poly(ethylene glycol).
Ultra-high mechanical properties of porous composites based on regenerated cellulose and cross-linked poly(ethylene glycol).
Carbohydrate polymers (2017-11-08)
Jian Teng, Biao Yang, Liang-Qing Zhang, Sheng-Qiang Lin, Ling Xu, Gan-Ji Zhong, Jian-Hua Tang, Zhong-Ming Li
ABSTRACT
The ultra-high mechanical, biocompatible and biodegradable porous regenerated cellulose/poly(ethylene glycol) (RC/PEG) composites with double network structure were fabricated via an simple method to dissolve cellulose followed by UV irradiation. The porous structure of RC/PEG was sensitively altered by PEG contents, which led to the porous structure morphology transition from 3D fibrillar network to close-grained sheet-like-network with the loading of cross-linked PEG. The porous RC/PEG showed excellent mechanical properties, i.e., the compressive strength can reach 33 times higher than that of neat RC (0.07MPa) at the compressive strain of 30%. Porous RC/PEG also displayed outstanding properties with openly porous structure and structural stabilization. Besides, porous RC/PEG exhibited good water absorbency, which the water absorbency ratio at equilibrium state was 83% higher than that of porous RC. This work provides an environmentally friendly and simple pathway to prepare non-toxic and biocompatible porous regenerated cellulose-based composites with high strength, structural stabilization and good water absorbency, which could be useful for packaging, biomedical applications, sewage purification, etc.
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Pricing and availability is not currently available.