Merck
  • Functional crackers: incorporation of the dietary fibers extracted from citrus seeds.
Functional crackers: incorporation of the dietary fibers extracted from citrus seeds.
Journal of food science and technology (2017-10-05)
Emin Yilmaz, Elif Karaman
ABSTRACT
Dietary fibers extracted from defatted press meals of orange and grapefruit seeds were used in the production of crackers and the crackers were evaluated for physicochemical, textural, sensory properties, and bioactive compounds. The unexplored fibers (grapes seeds and orange seeds) and wheat fiber were added at 2.9% levels. The effects of incorporation of different fibers on composition and consumer acceptability were evaluated. The textural stability during storage was also monitored. Although there were no significant differences for proximate compositions; fiber contents, antioxidant capacities, and phenolics compositions were significantly higher in the crackers containing orange seeds fiber. Sensory analysis showed that taste/flavor attribute scores were low, while appearance scores were equal to the control sample. The fracturability, hardness and water activity values remain constant up to 90 days storage at room temperature. The crackers containing orange seed and grapes seed fiber could provide some health benefits to consumers due to their high fiber and flavonoid contents.
Product Description
Supelco
Syringic acid, analytical standard
Pricing and availability is not currently available.