Merck
  • Inelastic neutron scattering study of reline: shedding light on the hydrogen bonding network of deep eutectic solvents.
Inelastic neutron scattering study of reline: shedding light on the hydrogen bonding network of deep eutectic solvents.
Physical chemistry chemical physics : PCCP (2017-07-01)
C F Araujo, J A P Coutinho, M M Nolasco, S F Parker, P J A Ribeiro-Claro, S Rudić, B I G Soares, P D Vaz
ABSTRACT
The solids choline chloride and urea, mixed in a 1 : 2 molar proportion, form the iconic deep eutectic solvent "Reline". A combination of computational and vibrational spectroscopy tools, including inelastic neutron scattering (INS), have been used to probe intermolecular interactions in the eutectic mixture. Reline's experimental spectra were estimated using discrete and periodic ab initio calculations of a molecular aggregate with two choline chloride and four urea units. This is the minimum size required to achieve satisfactory agreement with experiment, as smaller clusters cannot represent all of reline's significant intermolecular interactions. The INS spectrum of reline, compared with that of pure choline chloride, reveals a displacement of chloride anions away from their preferred positions on top of choline's methyl groups, whose torsional movement becomes less hindered in the mixture. Urea, which adopts a planar (sp
Sigma-Aldrich
Urea-d4, 98 atom % D
Pricing and availability is not currently available.