- Characterization and in vitro sensitivity of cholinesterases of gilthead seabream (Sparus aurata) to organophosphate pesticides.
Characterization and in vitro sensitivity of cholinesterases of gilthead seabream (Sparus aurata) to organophosphate pesticides.
Fish physiology and biochemistry (2016-10-08)
G Albendín, J M Arellano, M P Mánuel-Vez, C Sarasquete, M I Arufe
PMID27714546
ABSTRACT
The characterization of cholinesterase activity in brain and muscle of gilthead seabream was carried out using four specific substrates and three selective inhibitors. In addition, K