Merck
  • Exploring High-Performance n-Type Thermoelectric Composites Using Amino-Substituted Rylene Dimides and Carbon Nanotubes.
Exploring High-Performance n-Type Thermoelectric Composites Using Amino-Substituted Rylene Dimides and Carbon Nanotubes.
ACS nano (2017-05-17)
Guangbao Wu, Zhi-Guo Zhang, Yongfang Li, Caiyan Gao, Xin Wang, Guangming Chen
ABSTRACT
Taking advantage of the high electrical conductivity of a single-walled carbon nanotube (SWCNT) and the large Seebeck coefficient of rylene diimide, a convenient strategy is proposed to achieve high-performance n-type thermoelectric (TE) composites containing a SWCNT and amino-substituted perylene diimide (PDINE) or naphthalene diimide (NDINE). The obtained n-type composites display greatly enhanced TE performance with maximum power factors of 112 ± 8 (PDINE/SWCNT) and 135 ± 14 (NDINE/SWCNT) μW m
Sigma-Aldrich
1,4,5,8-Naphthalenetetracarboxylic dianhydride
Pricing and availability is not currently available.