Merck
  • Mass spectrometric analysis of intact human monoclonal antibody aggregates fractionated by size-exclusion chromatography.
Mass spectrometric analysis of intact human monoclonal antibody aggregates fractionated by size-exclusion chromatography.
Pharmaceutical research (2010-08-04)
Başak Kükrer, Vasco Filipe, Esther van Duijn, Piotr T Kasper, Rob J Vreeken, Albert J R Heck, Wim Jiskoot
ABSTRACT
The aim of this study was to develop a method to characterize intact soluble monoclonal IgG1 antibody (IgG) oligomers by mass spectrometry. IgG aggregates (dimers, trimers, tetramers and high-molecular-weight oligomers) were created by subjecting an IgG formulation to several pH jumps. Protein oligomer fractions were isolated by high performance size exclusion chromatography (HP-SEC), dialyzed against ammonium acetate pH 6.0 (a mass spectrometry-compatible volatile buffer), and analyzed by native electrospray ionization time-of-flight mass spectrometry (ESI-TOF MS). Monomeric and aggregated IgG fractions in the stressed IgG formulation were successfully isolated by HP-SEC. ESI-TOF MS analysis enabled us to determine the molecular weight of the monomeric IgG as well as the aggregates, including dimers, trimers and tetramers. HP-SEC separation and sample preparation proved to be necessary for good quality signal in ESI-TOF MS. Both the HP-SEC protocol and the ESI-TOF mass spectrometric technique were shown to leave the IgG oligomers largely intact. ESI-TOF MS is a useful tool complementary to HP-SEC to identify and characterize small oligomeric protein aggregates.
Product Description
Supelco
Cesium iodide, analytical standard, suitable for mass spectrometry (MS)
Pricing and availability is not currently available.