Merck
  • Design and Synthesis of Galactosylated Bifurcated Ligands with Nanomolar Affinity for Lectin LecA from Pseudomonas aeruginosa.
Design and Synthesis of Galactosylated Bifurcated Ligands with Nanomolar Affinity for Lectin LecA from Pseudomonas aeruginosa.
Chembiochem : a European journal of chemical biology (2017-03-21)
Anthony Angeli, Muchen Li, Lucie Dupin, Gérard Vergoten, Mathieu Noël, Mimouna Madaoui, Shuai Wang, Albert Meyer, Thomas Géhin, Sébastien Vidal, Jean-Jacques Vasseur, Yann Chevolot, François Morvan
ABSTRACT
Lectin A (LecA) from Pseudomonas aeruginosa is an established virulence factor. Glycoclusters that target LecA and are able to compete with human glycoconjugates present on epithelial cells are promising candidates to treat P. aeruginosa infection. A family of 32 glycodendrimers of generation 0 and 1 based on a bifurcated bis-galactoside motif have been designed to interact with LecA. The influences both of the central multivalent core and of the aglycon of these glycodendrimers on their affinity toward LecA have been evaluated by use of a microarray technique, both qualitatively for rapid screening of the binding properties and also quantitatively (K
Sigma-Aldrich
3-Azido-1-propanamine, ≥95%
Pricing and availability is not currently available.