Merck
  • A generalised individual-based algorithm for modelling the evolution of quantitative herbicide resistance in arable weed populations.
A generalised individual-based algorithm for modelling the evolution of quantitative herbicide resistance in arable weed populations.
Pest management science (2016-05-14)
Chun Liu, Melissa E Bridges, Shiv S Kaundun, Les Glasgow, Micheal Dk Owen, Paul Neve
ABSTRACT
Simulation models are useful tools for predicting and comparing the risk of herbicide resistance in weed populations under different management strategies. Most existing models assume a monogenic mechanism governing herbicide resistance evolution. However, growing evidence suggests that herbicide resistance is often inherited in a polygenic or quantitative fashion. Therefore, we constructed a generalised modelling framework to simulate the evolution of quantitative herbicide resistance in summer annual weeds. Real-field management parameters based on Amaranthus tuberculatus (Moq.) Sauer (syn. rudis) control with glyphosate and mesotrione in Midwestern US maize-soybean agroecosystems demonstrated that the model can represent evolved herbicide resistance in realistic timescales. Sensitivity analyses showed that genetic and management parameters were impactful on the rate of quantitative herbicide resistance evolution, whilst biological parameters such as emergence and seed bank mortality were less important. The simulation model provides a robust and widely applicable framework for predicting the evolution of quantitative herbicide resistance in summer annual weed populations. The sensitivity analyses identified weed characteristics that would favour herbicide resistance evolution, including high annual fecundity, large resistance phenotypic variance and pre-existing herbicide resistance. Implications for herbicide resistance management and potential use of the model are discussed. © 2016 Society of Chemical Industry.
Product Description
Supelco
Mesotrione, PESTANAL®, analytical standard
Pricing and availability is not currently available.