Merck
  • A selective ACAT-1 inhibitor, K-604, suppresses fatty streak lesions in fat-fed hamsters without affecting plasma cholesterol levels.
A selective ACAT-1 inhibitor, K-604, suppresses fatty streak lesions in fat-fed hamsters without affecting plasma cholesterol levels.
Atherosclerosis (2006-07-06)
Mami Ikenoya, Yasunobu Yoshinaka, Hideyuki Kobayashi, Katsumi Kawamine, Kimiyuki Shibuya, Fumiyasu Sato, Kimio Sawanobori, Takuya Watanabe, Akira Miyazaki
ABSTRACT
Acyl-coenzyme A:cholesterol O-acyltransferase-1 (ACAT-1), a major ACAT isozyme in macrophages, plays an essential role in foam cell formation in atherosclerotic lesions. However, whether pharmacological inhibition of macrophage ACAT-1 causes exacerbation or suppression of atherosclerosis is controversial. We developed and characterized a novel ACAT inhibitor, K-604. The IC(50) values of K-604 for human ACAT-1 and ACAT-2 were 0.45 and 102.85 micromol/L, respectively, indicating that K-604 is 229-fold more selective for ACAT-1. Kinetic analysis indicated that the inhibition was competitive with respect to oleoyl-coenzyme A with a K(i) value of 0.378 micromol/L. Exposure of human monocyte-derived macrophages to K-604 inhibited cholesterol esterification with IC(50) of 68.0 nmol/L. Furthermore, cholesterol efflux from THP-1 macrophages to HDL(3) or apolipoprotein A-I was enhanced by K-604. Interestingly, administration of K-604 to F1B hamsters on a high-fat diet at a dose of >or=1mg/kg suppressed fatty streak lesions without affecting plasma cholesterol levels. K-604, a potent and selective inhibitor of ACAT-1, suppressed the development of atherosclerosis in an animal model without affecting plasma cholesterol levels, providing direct evidence that pharmacological inhibition of ACAT-1 in the arterial walls leads to suppression of atherosclerosis.
Sigma-Aldrich
K-604, ≥98% (HPLC)
Pricing and availability is not currently available.