Merck
  • Pharmacodynamic and pharmacokinetic profiling of delafloxacin in a murine lung model against community-acquired respiratory tract pathogens.
Pharmacodynamic and pharmacokinetic profiling of delafloxacin in a murine lung model against community-acquired respiratory tract pathogens.
International journal of antimicrobial agents (2016-11-05)
Abrar K Thabit, Jared L Crandon, David P Nicolau
ABSTRACT
Increasing antimicrobial resistance in community-acquired pneumonia (CAP) pathogens has contributed to infection-related morbidity and mortality. Delafloxacin is a novel fluoroquinolone with broad-spectrum activity against Gram-positive and -negative organisms, including Streptococcus pneumoniae and methicillin-resistant Staphylococcus aureus (MRSA). This study aimed to define the pharmacodynamic profile of delafloxacin against CAP pathogens using a neutropenic murine lung infection model. Five S. pneumoniae, 2 methicillin-susceptible S. aureus (MSSA), 2 MRSA and 2 Klebsiella pneumoniae isolates were studied. Delafloxacin doses varied from 0.5 mg/kg/day to 640 mg/kg/day and were given as once-daily to every 3 h regimens over the 24-h treatment period. Efficacy was measured as the change in log10 CFU at 24 h compared with 0-h controls. Plasma and bronchopulmonary pharmacokinetic studies were conducted. Delafloxacin demonstrated potent in vitro and in vivo activity. Delafloxacin demonstrated high penetration into the lung compartment, as epithelial lining fluid concentrations were substantially higher than free drug in plasma. The ratio of the area under the free drug concentration-time curve to the minimum inhibitory concentration of the infecting organism (fAUC/MIC) was the parameter that best correlated with the efficacy of the drug, and the magnitude required to achieve 1 log10 CFU reduction was 31.8, 24.7, 0.4 and 9.6 for S. pneumoniae, MRSA, MSSA and K. pneumoniae, respectively. The observed in vivo efficacy of delafloxacin was supported by the high pulmonary disposition of the compound. The results derived from this pre-clinical lung model support the continued investigation of delafloxacin for the treatment of community-acquired lower respiratory tract infections.
Product Description
Supelco
Levofloxacin, analytical standard
Pricing and availability is not currently available.