Merck
  • In Vitro Activity of Delafloxacin Tested against Isolates of Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis.
In Vitro Activity of Delafloxacin Tested against Isolates of Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis.
Antimicrobial agents and chemotherapy (2016-07-28)
Robert K Flamm, Paul R Rhomberg, Michael D Huband, David J Farrell
ABSTRACT
Delafloxacin, an investigational anionic fluoroquinolone, is active against a broad range of Gram-positive and Gram-negative bacteria. In this study, 200 Streptococcus pneumoniae (plus 30 levofloxacin-resistant isolates), 200 Haemophilus influenzae, and 100 Moraxella catarrhalis isolates selected primarily from the United States (2014) were tested against delafloxacin and comparator agents. Delafloxacin was the most potent agent tested. MIC50 and MIC90 values against all S. pneumoniae isolates were 0.008 and 0.015 μg/ml. Delafloxacin susceptibility was not affected by β-lactamase status against H. influenzae and M. catarrhalis.
Sigma-Aldrich
Delafloxacin, ≥98% (HPLC)
Pricing and availability is not currently available.