Merck
  • Resin-capture and release strategy toward combinatorial libraries of 2,6,9-substituted purines.
Resin-capture and release strategy toward combinatorial libraries of 2,6,9-substituted purines.
Journal of combinatorial chemistry (2002-03-12)
Sheng Ding, Nathanael S Gray, Qiang Ding, Xu Wu, Peter G Schultz
ABSTRACT
A resin-capture and release strategy for making combinatorial 2,6,9-trisubstituted purine libraries is demonstrated by capturing N9-derivatized purines at the C6 position with a thio-modified polymer. The C2 fluoro group is subsequently substituted with primary and secondary amines followed by thioether oxidation and release by C6 substitution with amines and anilines. This approach complements a previously reported strategy where a 6-phenylsulfenylpurine scaffold was captured at the C2 position with a resin-bound amine.(3)
Sigma-Aldrich
2-Bromohypoxanthine, 96%
Pricing and availability is not currently available.