Merck
  • Studies on the synthesis of (-)-gymnodimine. Subunit synthesis and coupling.
Studies on the synthesis of (-)-gymnodimine. Subunit synthesis and coupling.
The Journal of organic chemistry (2007-02-01)
James D White, Laura Quaranta, Guoqiang Wang
ABSTRACT
Two principal subunits of the marine algal toxin (-)-gymnodimine were synthesized. A trisubstituted tetrahydrofuran representing C10-C18 of the toxin was prepared via a highly stereoselective iodine-mediated cyclization of an acyclic alkene bearing a bis-2,6-dichlorobenzyl (DCB) ether. The formation of a cis-2,5-disubstituted tetrahydrofuran in this process conforms to a stereodirecting effect by the DCB group proposed by Bartlett and Rychnovsky. A cyclohexene subunit corresponding to the C1-C8, C19-C24 portion of gymnodimine was synthesized via Diels-Alder cycloaddition of a 1,2,3-trisubstituted diene to a symmetrical dienophile obtained from Meldrum's acid. Differentiation of carbonyl groups in the cycloadduct was made by an intramolecular reaction with a neighboring alcohol to form a gamma-lactone. Linkage of the two subunits at C18-C19 was accomplished by using a B-alkyl Suzuki coupling in which a borane prepared from the pendent alkenyl chain of the cyclohexene domain was reacted with the (E)-iodoalkene attached at C16 of the tetrahydrofuran sector. Subsequent transformations positioned functional groups in the coupled product for a future macrocyclization event that would close the 15-membered ring of gymnodimine.
Sigma-Aldrich
(−)-Methyl (S)-2,2-dimethyl-1,3-dioxolane-4-carboxylate, 96%
Pricing and availability is not currently available.