Merck
  • Discovery of a new boron-containing antifungal agent, 5-fluoro-1,3-dihydro-1-hydroxy-2,1- benzoxaborole (AN2690), for the potential treatment of onychomycosis.
Discovery of a new boron-containing antifungal agent, 5-fluoro-1,3-dihydro-1-hydroxy-2,1- benzoxaborole (AN2690), for the potential treatment of onychomycosis.
Journal of medicinal chemistry (2006-07-21)
Stephen J Baker, Yong-Kang Zhang, Tsutomu Akama, Agnes Lau, Huchen Zhou, Vincent Hernandez, Weimin Mao, M R K Alley, Virginia Sanders, Jacob J Plattner
ABSTRACT
A structure-activity relationship investigation for a more efficacious therapy to treat onychomycosis, a fungal infection of the toe and fingernails, led to the discovery of a boron-containing small molecule, 5-fluoro-1,3-dihydro-1-hydroxy-2,1-benzoxaborole (AN2690), which is currently in clinical trials for onychomycosis topical treatment.
Sigma-Aldrich
2-Bromo-5-fluorobenzaldehyde, 96%
Pricing and availability is not currently available.