Merck
  • Improved tumor targeting and antitumor activity of camptothecin loaded solid lipid nanoparticles by preinjection of blank solid lipid nanoparticles.
Improved tumor targeting and antitumor activity of camptothecin loaded solid lipid nanoparticles by preinjection of blank solid lipid nanoparticles.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-05-03)
Dong-Jin Jang, Cheol Moon, Euichaul Oh
ABSTRACT
This study aimed to enhance the in vivo antitumor effects of camptothecin (CPT), a strong antitumor agent whose delivery is limited by poor aqueous solubility and instability of the active lactone form. CPT was loaded into sterically stabilized, solid lipid nanoparticles (CPT-SLNs) formulated for intravenous administration. The influence of preinjected blank SLNs on the tumor targeting, pharmacokinetics and antitumor activity of CPT-SLNs was investigated. The CPT-SLNs composed of trilaurin-based lipid matrix containing poloxamer188 and pegylated phospholipid as stabilizers were prepared by hot homogenization method and evaluated for in vitro characteristics and in vivo performance. The CPT-SLNs showed an in vitro long-term sustained release pattern and effectively protected the CPT lactone form from hydrolysis under physiological conditions. Notable tumor targeting and tumor growth inhibition were observed after intravenous administration of CPT-SLNs to mice with subcutaneous transplants of CT26 carcinoma cells. In pharmacokinetic studies in rats, CPT-SLNs markedly elevated plasma CPT level and prolonged blood circulation compared to free CPT. Nonetheless, high uptake of CPT-SLNs by reticuloendothelial system (RES)-rich tissues resulted in limited tumor targeting of CPT-SLNs and plasma CPT levels. Preinjection of blank SLNs before administration of CPT-SLNs to tumor-bearing mice substantially reduced the accumulation of CPT-SLNs in RES organs. This led to significantly enhanced tumor targeting, improved pharmacokinetic parameters and increased antitumor efficacy of CPT-SLNs. These results suggested that the in vivo antitumor effects of CPT-SLNs could be further enhanced by preinjection of blank SLNs. Therefore, CPT-SLNs with preinjected blank SLNs could be a potential approach for stable and effective CPT-based cancer therapy.
Product Description
Trilaurin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.