Merck
  • AtOPR3 specifically inhibits primary root growth in Arabidopsis under phosphate deficiency.
AtOPR3 specifically inhibits primary root growth in Arabidopsis under phosphate deficiency.
Scientific reports (2016-04-23)
Hongyan Zheng, Xiaoying Pan, Yuxia Deng, Huamao Wu, Pei Liu, Xuexian Li
ABSTRACT
The primary root plays essential roles in root development, nutrient absorption, and root architectural establishment. Primary root growth is generally suppressed by phosphate (P) deficiency in A. thaliana; however, the underlying molecular mechanisms are largely elusive to date. We found that AtOPR3 specifically inhibited primary root growth under P deficiency via suppressing root tip growth at the transcriptional level, revealing an important novel function of AtOPR3 in regulating primary root response to the nutrient stress. Importantly, AtOPR3 functioned to down-regulate primary root growth under P limitation mostly by its own, rather than depending on the Jasmonic acid signaling pathway. Further, AtOPR3 interacted with ethylene and gibberellin signaling pathways to regulate primary root growth upon P deficiency. In addition, the AtOPR3's function in inhibiting primary root growth upon P limitation was also partially dependent on auxin polar transport. Together, our studies provide new insights into how AtOPR3, together with hormone signaling interactions, modulates primary root growth in coping with the environmental stress in Arabidopsis.
Sigma-Aldrich
Ancymidol, suitable for plant cell culture, BioReagent
Pricing and availability is not currently available.