Merck
Cobalt-catalyzed aryl-sulfur bond formation.
Organic letters (2006-11-17)
Ying-Chieh Wong, Thiruvellore Thatai Jayanth, Chien-Hong Cheng
ABSTRACT
A new cobalt-catalyzed coupling of aryl halides with thiophenols and alkanethiols is reported. A variety of aryl sulfides can be prepared in excellent yields under mild reaction conditions using 1-2 mol % of CoI2(dppe) and Zn. This new cobalt-catalyzed coupling represents an interesting addition to previously known methods to synthesize thioethers. [reaction: see text].
Sigma-Aldrich
4-Iodotoluene, 99%
Pricing and availability is not currently available.