Merck
Synthesis of 3-fluoroazetidines.
The Journal of organic chemistry (2006-08-26)
Willem Van Brabandt, Guido Verniest, Dirk De Smaele, Guillaume Duvey, Norbert De Kimpe
ABSTRACT
N-(Alkylidene or 1-arylmethylidene)-2-propenylamines were regiospecifically functionalized to novel N-(alkylidene or 1-arylmethylidene)-3-bromo-2-fluoropropylamines, which were proven to be excellent precursors for 3-fluoroazetidines.
Sigma-Aldrich
Triethylamine trihydrofluoride, 97%
Pricing and availability is not currently available.