Merck
  • Multi-centre evaluation of real-time multiplex PCR for detection of carbapenemase genes OXA-48, VIM, IMP, NDM and KPC.
Multi-centre evaluation of real-time multiplex PCR for detection of carbapenemase genes OXA-48, VIM, IMP, NDM and KPC.
BMC infectious diseases (2014-01-16)
Anneke van der Zee, Lieuwe Roorda, Gerda Bosman, Ad C Fluit, Mirjam Hermans, Paul H M Smits, Adri G M van der Zanden, René Te Witt, Lesla E S Bruijnesteijn van Coppenraet, James Cohen Stuart, Jacobus M Ossewaarde
ABSTRACT
Resistance to carbapenem antibiotics is emerging worldwide among Enterobacteriaceae. To prevent hospital transmission due to unnoticed carriage of carbapenemase producing micro-organisms in newly admitted patients, or follow-up of patients in an outbreak setting, a molecular screening method was developed for detection of the most prevalent carbapenemase genes; blaOXA-48, blaVIM, blaIMP, blaNDM and blaKPC. A real-time multiplex PCR assay was evaluated using a collection of 86 Gram negative isolates, including 62 carbapenemase producers. Seven different laboratories carried out this method and used the assay for detection of the carbapenemase genes on a selection of 20 isolates. Both sensitivity and specificity of the multiplex PCR assay was 100%, as established by results on the strain collection and the inter-laboratory comparisons. In this study, we present a multiplex real-time PCR that is a robust, reliable and rapid method for the detection of the most prevalent carbapenemases blaOXA-48, blaVIM, blaIMP, blaNDM and blaKPC, and is suitable for screening of broth cultured rectal swabs and for identification of carbapenemase genes in cultures.
Sigma-Aldrich
Dilution Solution, Extract-N-Amp Plant Dilution Solution
Pricing and availability is not currently available.