Merck
  • SimplePHOX, a readily available chiral ligand system for iridium-catalyzed asymmetric hydrogenation.
SimplePHOX, a readily available chiral ligand system for iridium-catalyzed asymmetric hydrogenation.
Organic letters (2004-06-05)
Sebastian P Smidt, Frederik Menges, Andreas Pfaltz
ABSTRACT
[reaction: see text] New Ir-SimplePHOX complexes Ir-6-Ir-9 catalyze the quantitative, highly enantioselective hydrogenation of a range of unfunctionalized and functionalized olefins. Synthesis, catalytic results, and X-ray crystal structures are presented here.
Sigma-Aldrich
L-tert-Leucinol, 98%
Pricing and availability is not currently available.