Merck
  • Nanotherapeutics in angiogenesis: synthesis and in vivo assessment of drug efficacy and biocompatibility in zebrafish embryos.
Nanotherapeutics in angiogenesis: synthesis and in vivo assessment of drug efficacy and biocompatibility in zebrafish embryos.
International journal of nanomedicine (2011-10-07)
Jinping Cheng, Yan-Juan Gu, Yajun Wang, Shuk Han Cheng, Wing-Tak Wong
ABSTRACT
Carbon nanotubes have shown broad potential in biomedical applications, given their unique mechanical, optical, and chemical properties. In this pilot study, carbon nanotubes have been explored as multimodal drug delivery vectors that facilitate antiangiogenic therapy in zebrafish embryos. Three different agents, ie, an antiangiogenic binding site (cyclic arginine-glycin-easpartic acid), an antiangiogenic drug (thalidomide), and a tracking dye (rhodamine), were conjugated onto single-walled carbon nanotubes (SWCNT). The biodistribution, efficacy, and biocompatibility of these triple functionalized SWCNT were tested in mammalian cells and validated in transparent zebrafish embryos. Accumulation of SWCNT-associated nanoconjugates in blastoderm cells facilitated drug delivery applications. Mammalian cell xenograft assays demonstrated that these antiangiogenic SWCNT nanoconjugates specifically inhibited ectopic angiogenesis in the engrafted zebrafish embryos. This study highlights the potential of using SWCNT for generating efficient nanotherapeutics.
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
2-(Boc-amino)ethyl bromide, ≥97.0% (GC)
Pricing and availability is not currently available.