Merck
  • Efficient red luminescence from organic-soluble Au₂₅ clusters by ligand structure modification.
Efficient red luminescence from organic-soluble Au₂₅ clusters by ligand structure modification.
Nanoscale (2015-08-06)
Ammu Mathew, Elizabeth Varghese, Susobhan Choudhury, Samir Kumar Pal, T Pradeep
ABSTRACT
An efficient method to enhance visible luminescence in a visibly non-luminescent organic-soluble 4-(tert butyl)benzyl mercaptan (SBB)-stabilized Au25 cluster has been developed. This method relies mainly on enhancing the surface charge density on the cluster by creating an additional shell of thiolate on the cluster surface, which enhances visible luminescence. The viability of this method has been demonstrated by imparting red luminescence to various ligand-protected quantum clusters (QCs), observable to the naked eye. The bright red luminescent material derived from Au25SBB18 clusters was characterized using UV-vis and luminescence spectroscopy, TEM, SEM/EDS, XPS, TG, ESI and MALDI mass spectrometry, which collectively proposed an uncommon molecular formula of Au29SBB24S, suggested to be due to different stapler motifs protecting the Au25 core. The critical role of temperature on the emergence of luminescence in QCs has been studied. The restoration of the surface ligand shell on the Au25 cluster and subsequent physicochemical modification to the cluster were probed by various mass spectral and spectroscopic techniques. Our results provide fundamental insights into the ligand characteristics determining luminescence in QCs.
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Pricing and availability is not currently available.
Sigma-Aldrich
2-Phenylethanethiol, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylethyl mercaptan, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
VenPure® SF, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-tert-Butylbenzyl mercaptan
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Pricing and availability is not currently available.
Tantalum(V) ethoxide, packaged for use in deposition systems
Pricing and availability is not currently available.