Merck
  • Nanographene oxide-hyaluronic acid conjugate for photothermal ablation therapy of skin cancer.
Nanographene oxide-hyaluronic acid conjugate for photothermal ablation therapy of skin cancer.
ACS nano (2014-01-05)
Ho Sang DJung, Won Ho Kong, Dong Kyung Sung, Min-Young Lee, Song Eun Beack, Do Hee Keum, Ki Su Kim, Seok Hyun Yun, Sei Kwang Hahn
ABSTRACT
Melanoma skin cancer is one of the most dangerous skin cancers and the main cause of skin-cancer-related mortality. Hyaluronic acid (HA) has been used as an effective transdermal delivery carrier of chemical drugs and biopharmaceuticals. In this work, a nanographene oxide-HA conjugate (NGO-HA) was synthesized for photothermal ablation therapy of melanoma skin cancer using a near-infrared (NIR) laser. Confocal microscopy and ex vivo bioimaging clearly visualized the remarkable transdermal delivery of NGO-HA to tumor tissues in the skin of mice, which might be ascribed to highly expressed HA receptors and relatively leaky structures around tumor tissues, enabling the enhanced permeation and retention of nanoparticles. The NIR irradiation resulted in complete ablation of tumor tissues with no recurrence of tumorigenesis. The antitumor effect was confirmed by ELISA for caspase-3 activity and histological and immunohistochemical analyses with TUNEL assay for tumor apoptosis. Taken together, we could confirm the feasibility of transdermal NGO-HA for photothermal ablation therapy of melanoma skin cancers.
Sigma-Aldrich
Graphene oxide nanocolloids, 2 mg/mL, dispersion in H2O
Pricing and availability is not currently available.