Merck
  • Fluorine-functionalized and simultaneously reduced graphene oxide as a novel hole transporting layer for highly efficient and stable organic photovoltaic cells.
Fluorine-functionalized and simultaneously reduced graphene oxide as a novel hole transporting layer for highly efficient and stable organic photovoltaic cells.
Nanoscale (2014-05-08)
Su-Hyeon Kim, Cheol-Ho Lee, Jin-Mun Yun, Yong-Jin Noh, Seok-Soon Kim, Sungho Lee, Seung Mu Jo, Han-Ik Joh, Seok-In Na
ABSTRACT
A one-step reduction and functionalization of graphene oxide (FrGO) was easily achieved using a novel phenylhydrazine-based reductant containing fluorine atoms, which can induce p-type doping due to its high electronegativity. The FrGO-based OPV exhibited a high power conversion efficiency of ∼6.71% and a superior OPV-stability to commercial PEDOT:PSS.
Sigma-Aldrich
4-(Trifluoromethyl)phenylhydrazine, 96%
Pricing and availability is not currently available.