Merck
Switchable-polarity solvents prepared with a single liquid component.
The Journal of organic chemistry (2007-12-11)
Lam Phan, Jeremy R Andreatta, Loel K Horvey, Colin F Edie, Aimée-Lee Luco, Anish Mirchandani, Donald J Darensbourg, Philip G Jessop
ABSTRACT
Known liquids that can reversibly switch their polarity at atmospheric pressure are all prepared as mixtures of two liquid components; we now report a series of switchable-polarity solvents that consist, in their low-polarity form, of only a single liquid component, a secondary amine. These solvents operate in a polarity range that is significantly lower than those of previously reported switchable solvents. Application to the separation and purification of a polymer and recovery of a catalyst is described.
Sigma-Aldrich
Dimethylammonium dimethylcarbamate
Pricing and availability is not currently available.