Merck
6-Methyl-1,3,5-triazine-2,4-diamine butane-1,4-diol monosolvate.
Acta crystallographica. Section E, Structure reports online (2013-03-12)
Rajni M Bhardwaj, Iain Oswald, Alastair J Florence
ABSTRACT
The title co-crystal, C4H7N5·C4H10O2, crystallizes with one mol-ecule of 6-methyl-1,3,5-triazine-2,4-diamine (DMT) and one mol-ecule of butane-1,4-diol in the asymmetric unit. The DMT mol-ecules form ribbons involving centrosymmetric R2(2)(8) dimer motifs between DMT mol-ecules along the c-axis direction. These ribbons are further hydrogen bonded to each other through butane-1,4-diol, forming sheets parallel to (121).
Sigma-Aldrich
6-Methyl-1,3,5-triazine-2,4-diamine, 98%
Pricing and availability is not currently available.