Merck
  • Immunophenotype of high-grade prostatic adenocarcinoma and urothelial carcinoma.
Immunophenotype of high-grade prostatic adenocarcinoma and urothelial carcinoma.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2000-12-06)
E M Genega, B Hutchinson, V E Reuter, P B Gaudin
ABSTRACT
Morphologic features alone can usually be used to distinguish prostatic adenocarcinoma and urothelial carcinoma of the urinary bladder. Poorly differentiated tumors, however, can occasionally have features of both neoplasms, making determination of site of origin difficult. No study has provided a panel of antibodies to assist in the distinction of these two tumors. For this study, 73 examples of moderately and poorly differentiated prostatic adenocarcinoma and 46 examples of high-grade urothelial carcinoma were obtained from radical resection specimens. Immunohistochemical studies were performed using the following panel of antibodies: cytokeratin (CK) 7, CK 20, 34betaE12, Leu M1, carcinoembryonic antigen (CEA)m, CEAp, p53, Leu 7, prostate-specific acid phosphatase (PSAP), prostate-specific antigen (PSA), and B72.3. Mucicarmine was also performed. Intermediate and high-grade prostatic carcinoma were compared and then high-grade prostatic carcinoma was compared with high-grade urothelial carcinoma. PSA and PSAP each stained 94% of prostatic adenocarcinomas, but no urothelial carcinomas. Leu 7 stained 94% of prostate and 17% of urothelial carcinomas. Over half of the urothelial carcinomas showed positivity for 34betaE12 (65%), as did two cases of prostatic carcinoma (6%). Eighty-three percent of urothelial carcinomas and 12% of prostatic adenocarcinomas stained with CK 7. Forty-one percent of urothelial carcinomas and 12% of prostatic carcinomas were reactive for CEAm, and p53 stained 33% and 3% of urothelial and prostatic adenocarcinomas, respectively. No significant difference was seen in the expression of CEAp, CK 20, B72.3, Leu M1, or mucicarmine between prostate and urothelial carcinoma. We propose a panel of six antibodies to assist in the distinction of high-grade prostatic adenocarcinoma from high grade urothelial carcinoma: PSA, PSAP, 34betaE12, Leu 7, CK 7, and p53. The first three antibodies should be used initially; if results are negative, the remaining antibodies may be employed.
Sigma-Aldrich
PSAP (PASE/4LJ) Mouse Monoclonal Antibody
Pricing and availability is not currently available.