Merck
Neutrophil-rich anaplastic large cell lymphoma presenting in the skin.
The American Journal of dermatopathology (2003-03-26)
Naoko Kato, Osamu Mizuno, Kei Ito, Kumiko Kimura, Masahiko Shibata
ABSTRACT
A neutrophil-rich anaplastic large cell lymphoma (ALCL) presented in the skin of a 47-year-old Japanese woman. The solitary cutaneous lesion was an eroded, 10-mm, dome-shaped nodule involving the skin of her left upper arm. Histologically, it showed a proliferation of pleomorphic, anaplastic, large tumor cells with nuclei of various shapes, including embryo-shaped, Reed-Sternberg cell-like binucleated, and wreath-shaped multiple nuclei, in the dermis and subcutaneous tissues. There was an admixture of neutrophils ranging from about 30% to more than 50% per field. Immunophenotypically, the neoplastic cells were positive for CD30, CD4, leukocyte common antigen, anaplastic lymphoma kinase-1, epithelial membrane antigen, and granzyme B. She had no peripheral neutrophilia. The lesson was excised, and the site on the left upper arm was irradiated. Six and a half months after diagnosis, however, swelling of a left axillary lymph node appeared; it also showed a proliferation of anaplastic large tumor cells admixed with numerous neutrophils ranging from about 25% to more than 60% per field. Southern blot analysis of T-cell receptor gene rearrangement revealed a clonal band. The patient was treated with six courses of cyclophosphamide, doxorubicin, vincristine, and prednisolone chemotherapy with complete remission. Seventeen cases of neutrophil-rich ALCL arising in the skin, lymph node, muscle, testis, and skull bone were reviewed. This form may possibly be induced by trauma or irritation of conventional ALCL, although the true mechanism for the infiltration of neutrophils is still unclear.
Sigma-Aldrich
Granzyme B Rabbit Polyclonal Antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Granzyme B (EP230) Rabbit Monoclonal Primary Antibody
Pricing and availability is not currently available.