Merck
  • Convergence of the adhesive and fibrinolytic systems: recognition of urokinase by integrin alpha Mbeta 2 as well as by the urokinase receptor regulates cell adhesion and migration.
Convergence of the adhesive and fibrinolytic systems: recognition of urokinase by integrin alpha Mbeta 2 as well as by the urokinase receptor regulates cell adhesion and migration.
Blood (2002-10-24)
Elzbieta Pluskota, Dmitry A Soloviev, Edward F Plow
ABSTRACT
Previous studies demonstrated that integrin alpha(M)beta(2) (CD11b/18, Mac-1) forms a physical complex with the urokinase-type plasminogen activator receptor (uPAR/CD87) on leukocytes. In this study, we used human peripheral blood neutrophils and transfected cells expressing alpha(M)beta(2), uPAR, or both receptors to show that the integrin can directly interact with urokinase (uPA). We demonstrate that alpha(M)beta(2) supported adhesion and migration of these cells to uPA, and, in each case, blockade of alpha(M)beta(2) suppressed the response. Within uPA, both the kringle and proteolytic domains are recognized by alpha(M)beta(2), which are distinct from the growth factor domain that binds to uPAR. Within the alpha(M) subunit of the integrin, the I domain interacts with uPA, which is distinct from the region that interacts with uPAR. On cells expressing uPAR and alpha(M)beta(2), both receptors mediated adhesion and migration. This cooperation was particularly apparent in the responses of neutrophils to uPA, where blockade of alpha(M)beta(2) reduced uPAR-mediated responses and engagement of uPAR enhanced recognition of uPA by alpha(M)beta(2). Thus, recognition of uPA by alpha(M)beta(2) allows for formation of a multicontact trimolecular complex, in which a single uPA ligand may bind simultaneously to both uPAR and alpha(M)beta(2). This complex may play an important role in the control of inflammatory cell migration and vascular homeostasis.
Sigma-Aldrich
Anti-Integrin β2 Antibody, clone P4H9, clone P4H9, Chemicon®, from mouse
Pricing and availability is not currently available.