Merck
  • Validation of a reversed-phase HPLC method for quantitative amino acid analysis.
Validation of a reversed-phase HPLC method for quantitative amino acid analysis.
Journal of biomolecular techniques : JBT (2006-06-03)
Maria Paola Bartolomeo, Federico Maisano
ABSTRACT
A semi-automated method for amino acid derivatization and analysis has been validated for use in analysis of protein biopharmaceuticals. The method includes protein hydrolysis, o-phthalaldehyde derivatization, and reversed-phase high-performance liquid chromatography analysis in a general-purpose UV-visible high-performance liquid chromatography system. Amino-acid derivatization is performed automatically by the high-performance liquid chromatography autosampler right before injection. The required validation parameters, i.e., specificity, linearity, accuracy, precision, limit of detection, and limit of quantification, were studied for bovine serum albumin and for a recombinant human Fab fragment. The method can be employed as an absolute quantification method for determination of extinction coefficients of recombinant proteins.
Product Description
Supelco
Insert for 2 mL standard opening vial, 4.6 mm I.D., volume 0.15 mL, conical clear glass (with bottom spring), O.D. × H 5 mm × 30 mm, pkg of 100 ea
Pricing and availability is not currently available.
Supelco
Insert for 2 mL standard opening vial, 4.6 mm I.D., volume 0.15 mL, conical polypropylene (with bottom spring), O.D. × H 5 mm × 30 mm, pkg of 100 ea
Pricing and availability is not currently available.
Supelco
Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.2 mL, conical clear glass (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
Pricing and availability is not currently available.