Merck
  • Hydrogen-bonding patterns in the cocrystal 2-amino-4,6-dimethoxy-pyrimidine-anthranilic acid (1/1).
Hydrogen-bonding patterns in the cocrystal 2-amino-4,6-dimethoxy-pyrimidine-anthranilic acid (1/1).
Acta crystallographica. Section E, Structure reports online (2007-01-01)
Kaliyaperumal Thanigaimani, Packianathan Thomas Muthiah, Daniel E Lynch
ABSTRACT
In the title cocrystal, C(6)H(9)N(3)O(2)·C(7)H(7)NO(2), the asymmetric unit contains two crystallographically independent 2-amino-4,6-dimeth-oxy pyrimidine-anthranilic acid adducts. The 2-amino-4,6-dimeth-oxy pyrimidine mol-ecules inter-act with the carboxylic group of the respective anthranilic acid mol-ecules through N-H⋯O and O-H⋯N hydrogen bonds, forming a cyclic hydrogen-bonded motif R(2) (2)(8). The pyrimidine mol-ecules also form base pairs via a pair of N-H⋯N hydrogen bonds, forming another R(2) (2)(8) motif. The typical intra-molecular N-H⋯O hydrogen bond is observed in the anthranilic acid mol-ecules. Furthermore, the crystal structure is stabilized by C-H⋯O hydrogen bonds.
Sigma-Aldrich
2-Amino-4,6-dimethoxypyrimidine, 98%
Pricing and availability is not currently available.