Merck
  • Carbon monoxide free aminocarbonylation of aryl and alkenyl iodides using DMF as an amide source.
Carbon monoxide free aminocarbonylation of aryl and alkenyl iodides using DMF as an amide source.
Organic letters (2002-08-17)
Kazushi Hosoi, Kyoko Nozaki, Tamejiro Hiyama
ABSTRACT
[reaction: see text] Palladium-catalyzed coupling reaction of N,N-dimethylformamide with aryl or alkenyl halides successfully proceeded in the presence of phosphoryl chloride to afford the corresponding tertiary amides in good yields.
Sigma-Aldrich
4-Iodobenzotrifluoride, 97%
Pricing and availability is not currently available.