Merck
  • Predation and control efficacies of Misgurnus mizolepis (Cypriniformes: Cobitidae) toward Culex pipiens molestus (Diptera: Culicidae) and fish toxicity of temephos in laboratory and septic tank conditions.
Predation and control efficacies of Misgurnus mizolepis (Cypriniformes: Cobitidae) toward Culex pipiens molestus (Diptera: Culicidae) and fish toxicity of temephos in laboratory and septic tank conditions.
Journal of medical entomology (2014-08-15)
Seong Chun Chae, Young Hyun Kwon, Kyung Il Min, Hyung Soo Kim, Nam-Jin Kim, Jun-Ran Kim, Bong Gi Son, Young-Joon Ahn
ABSTRACT
Culex pipiens molestus Forskal (Diptera: Culicidae) is the dominant mosquito species in septic tanks in South Korea. An assessment was made of the biological control potential of mud loaches, Misgurnus mizolepis Günther (Cypriniformes: Cobitidae), toward Cx. p. molestus larvae in laboratory and septic tanks. Results were compared with those of temephos 20% emulsifiable concentrate. In laboratory tests, all mud loaches survived on sedimentation chamber- and effluent chamber-collected water of aerobic septic tanks (ASTs), whereas all mud loaches died within 3-12 h after introduction into sedimentation chamber- and effluent chamber-collected water of anaerobic septic tanks, Gill hyperplasia and hemorrhages at the bases of pectoral fins were detected in all dead mud loaches. These appeared to have been caused by bacterial disease, rather than the physical and chemical characteristics of the septic tank water. A mud loach consumed an average range of 1,072-1,058 larvae of Cx. p. molestus in the AST water at 24 h. At the manufacturer's recommended rate (10 ml/ton) in the AST water, the temephos formulation did not cause fish mortality. In the AST experiment, predation of mosquito larvae by mud loaches at a release rate of one fish per 900 mosquito larvae resulted in complete mosquito control from the third day after treatment throughout the 18-wk survey period, compared with temephos 20% emulsifiable concentrate-treated AST water (reduction rate, 40% at 28 days after treatment). Reasonable mosquito control in aerobic septic tanks can be achieved by mosquito breeding season stocking of a rate of one mud loach per 900 mosquito larvae.
Product Description
Supelco
Temephos, PESTANAL®, analytical standard
Pricing and availability is not currently available.