Merck
  • Synthesis of 9Z-9-substituted retinoic acids by palladium catalyzed coupling reaction of a vinyl triflate with alkenyl stannanes.
Synthesis of 9Z-9-substituted retinoic acids by palladium catalyzed coupling reaction of a vinyl triflate with alkenyl stannanes.
Chemical & pharmaceutical bulletin (2000-09-19)
A Wada, Y Nomoto, K Tano, E Yamashita, M Ito
ABSTRACT
Palladium catalyzed cross coupling reactions of a vinyl triflate intermediate and various alkenyl stannanes afforded trisubstituted Z-olefins stereoselectively in high yields. These olefins were then converted to the corresponding 9Z-retinoic acids via Horner-Emmons reaction and subsequent basic hydrolysis in excellent yields.
Sigma-Aldrich
2-Pentyn-1-ol, 98%
Pricing and availability is not currently available.